Acetone, 99+%, extra pure
Acetone, 99+%, extra pure
Acetone, 99+%, extra pure
Thermo Scientific Chemicals

Acetone, 99+%, extra pure

CAS: 67-64-1 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
View all
Plastic Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 177170250
Price (EUR)
392,00
Each
Quantity:
25 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (EUR)
392,00
Each
Chemical Identifiers
CAS67-64-1
IUPAC Namepropan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Water=<0.5 % (K.F.)
Residue after evaporation=<0.005 %
Appearance (Form)Clear liquid
Color scale=<10 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only