Iridium(III) chloride trihydrate, 53-56% Ir
Iridium(III) chloride trihydrate, 53-56% Ir
Iridium(III) chloride trihydrate, 53-56% Ir
Thermo Scientific Chemicals

Iridium(III) chloride trihydrate, 53-56% Ir

CAS: 13569-57-8 | Cl3Ir·3H2O | 352.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 195500050
Price (EUR)
2 164,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
2 164,00
Each
Chemical Identifiers
CAS13569-57-8
IUPAC Nametrichloroiridium trihydrate
Molecular FormulaCl3H6IrO3
InChI KeyLNJXVUXPFZKMNF-UHFFFAOYSA-K
SMILESO.O.O.Cl[Ir](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to black
Appearance (Form)Crystals
Assay53 to 56 % (Ir)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only