Potassium hexacyanocobaltate(III), 95%
Potassium hexacyanocobaltate(III), 95%
Potassium hexacyanocobaltate(III), 95%
Thermo Scientific Chemicals

Potassium hexacyanocobaltate(III), 95%

CAS: 13963-58-1 | C6CoK3N6 | 332.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 203921000
Price (EUR)
117,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
117,00
Each
Chemical Identifiers
CAS13963-58-1
IUPAC Namecobalt(3+) tripotassium hexakis(iminomethanide)
Molecular FormulaC6CoK3N6
InChI KeyLGRDAQPMSDIUQJ-UHFFFAOYSA-N
SMILES[K+].[K+].[K+].[Co+3].[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N.[C-]#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=94 %
Appearance (Form)Crystalline solid
Appearance (Color)Light yellow to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only