Glycine, sodium salt, 99%
Glycine, sodium salt, 99%
Glycine, sodium salt, 99%
Thermo Scientific Chemicals

Glycine, sodium salt, 99%

CAS: 6000-44-8 | C2H4NNaO2 | 97.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 204595000
Price (EUR)
257,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
257,00
Each
Chemical Identifiers
CAS6000-44-8
SpecificationsSpecification SheetSpecification Sheet
UV transmittance>=94.0 % (c=5, H2O, 400 nm) 1 cm cell
Appearance (Form)Powder or crystals
Appearance (Color)Colorless to white to off-white
Infrared spectrumConforms
Water=<1.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only