5-Hexen-1-ol, 99%
5-Hexen-1-ol, 99%
5-Hexen-1-ol, 99%
Thermo Scientific Chemicals

5-Hexen-1-ol, 99%

CAS: 821-41-0 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 213040050
Price (EUR)
82,10
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
82,10
Each
Chemical Identifiers
CAS821-41-0
IUPAC Namehex-5-en-1-ol
Molecular FormulaC6H12O
InChI KeyUIZVMOZAXAMASY-UHFFFAOYSA-N
SMILESOCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4340 to 1.4360 (20°C, 589 nm)
GC>=98.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only