1-Hydroxyethylethoxypiperazine, 95%
1-Hydroxyethylethoxypiperazine, 95%
1-Hydroxyethylethoxypiperazine, 95%
Thermo Scientific Chemicals

1-Hydroxyethylethoxypiperazine, 95%

CAS: 13349-82-1 | C8H18N2O2 | 174.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 259110250
Price (EUR)
147,00
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
147,00
Each
Chemical Identifiers
CAS13349-82-1
IUPAC Name4-[2-(2-hydroxyethoxy)ethyl]piperazine-1,4-diium
Molecular FormulaC8H20N2O2
InChI KeyFLNQAPQQAZVRDA-UHFFFAOYSA-P
SMILESOCCOCC[NH+]1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
Appearance (Color)Clear colorless to yellow-brown
Infrared spectrumConforms
Titration Tertiary base>=94 %
Titration with HClO4>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only