N-Carbobenzyloxy-L-valine, 99+%
N-Carbobenzyloxy-L-valine, 99+%
N-Carbobenzyloxy-L-valine, 99+%
Thermo Scientific Chemicals

N-Carbobenzyloxy-L-valine, 99+%

CAS: 1149-26-4 | C13H17NO4 | 251.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 275940250
Price (EUR)
91,70
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
91,70
Each
Chemical Identifiers
CAS1149-26-4
IUPAC Name(2S)-2-{[(benzyloxy)carbonyl]amino}-3-methylbutanoic acid
Molecular FormulaC13H17NO4
InChI KeyCANZBRDGRHNSGZ-NSHDSACASA-N
SMILESCC(C)[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=99.0 %
Water=<0.5 % (K.F.)
Appearance (Form)Crystalline powder
Specific optical rotation-3.2° to -4.8° (20°C, 589 nm) (c=2, acetic acid)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only