4-Chloro-2-fluorobenzaldehyde, 99%
4-Chloro-2-fluorobenzaldehyde, 99%
4-Chloro-2-fluorobenzaldehyde, 99%
Thermo Scientific Chemicals

4-Chloro-2-fluorobenzaldehyde, 99%

CAS: 61072-56-8 | C7H4ClFO | 158.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 290620050
Price (EUR)
98,40
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
98,40
Each
Chemical Identifiers
CAS61072-56-8
IUPAC Name4-chloro-2-fluorobenzaldehyde
Molecular FormulaC7H4ClFO
InChI KeyUVGYSEIWAOOIJR-UHFFFAOYSA-N
SMILESFC1=CC(Cl)=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow to light beige
GC>=98.5 %
Appearance (Form)Powder or crystals
Melting point57°C to 61°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only