Chloromethyl butyrate, 99%
Chloromethyl butyrate, 99%
Chloromethyl butyrate, 99%
Thermo Scientific Chemicals

Chloromethyl butyrate, 99%

CAS: 33657-49-7 | C5H9ClO2 | 136.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 295580250
Price (EUR)
116,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
116,00
Each
Chemical Identifiers
CAS33657-49-7
IUPAC Namechloromethyl butanoate
Molecular FormulaC5H9ClO2
InChI KeyBDPZFQLKFUONAG-UHFFFAOYSA-N
SMILESCCCC(=O)OCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only