L-Leucinamide hydrochloride, 99%
L-Leucinamide hydrochloride, 99%
L-Leucinamide hydrochloride, 99%
Thermo Scientific Chemicals

L-Leucinamide hydrochloride, 99%

CAS: 10466-61-2 | C6H15ClN2O | 166.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 302980010
Price (EUR)
24,20
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
24,20
Each
Chemical Identifiers
CAS10466-61-2
IUPAC Name(2S)-2-amino-4-methylpentanamide hydrochloride
Molecular FormulaC6H15ClN2O
InChI KeyVSPSRRBIXFUMOU-XZNNNFJINA-N
SMILESCl.CC(C)C[C@H](N)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Titration Argentometric>=98.5 %
Infrared spectrumConforms
Specific optical rotation+9° to +11° (20°C, 589 nm) (c=1, H2O)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only