Product Image
Thermo Scientific™

Thymoquinone

CAS: 490-91-5 | C10H12O2 | 164.20 g/mol
Have Questions?
Catalog NumberQuantity
3050700505 g
Catalog number 305070050
Price (EUR)
173,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
173,00
Each
Chemical Identifiers
CAS490-91-5
IUPAC Name2-methyl-5-(propan-2-yl)cyclohexa-2,5-diene-1,4-dione
Molecular FormulaC10H12O2
InChI KeyKEQHJBNSCLWCAE-UHFFFAOYSA-N
SMILESCC(C)C1=CC(=O)C(C)=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Yellow to orange to brown
Melting point44°C to 47°C
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only