2,2,2-Trifluoroethanesulfonyl chloride, 95%
2,2,2-Trifluoroethanesulfonyl chloride, 95%
2,2,2-Trifluoroethanesulfonyl chloride, 95%
Thermo Scientific Chemicals

2,2,2-Trifluoroethanesulfonyl chloride, 95%

CAS: 1648-99-3 | C2H2ClF3O2S | 182.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
2.5 g
10 g
Packaging:
Glass bottle
Catalog number 305350010
Price (EUR)
80,90
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
80,90
Each
Chemical Identifiers
CAS1648-99-3
IUPAC Name2,2,2-trifluoroethane-1-sulfonyl chloride
Molecular FormulaC2H2ClF3O2S
InChI KeyCXCHEKCRJQRVNG-UHFFFAOYSA-N
SMILESFC(F)(F)CS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Titration Argentometric>=94 % (ex Cl)
Infrared spectrumConforms
Refractive index1.3870 to 1.3890 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only