Indium(III) iodide, 99.998%, (trace metal basis)
Indium(III) iodide, 99.998%, (trace metal basis)
Indium(III) iodide, 99.998%, (trace metal basis)
Thermo Scientific Chemicals

Indium(III) iodide, 99.998%, (trace metal basis)

CAS: 13510-35-5 | InI3 | 495.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 316010050
Price (EUR)
168,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
168,00
Each
Chemical Identifiers
CAS13510-35-5
IUPAC Nameindium(3+) triiodide
Molecular FormulaI3In
InChI KeyRMUKCGUDVKEQPL-UHFFFAOYSA-K
SMILES[In+3].[I-].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric22.7 to 23.5 % (In)
Appearance (Form)Powder or crystals
Total trace metal impurities=<0.002 %
Appearance (Color)Yellow to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only