3-Isochromanone, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Isochromanone, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 370130010
Price (EUR)
28,70
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
28,70
Each
Chemical Identifiers
CAS4385-35-7
IUPAC Name3,4-dihydro-1H-2-benzopyran-3-one
Molecular FormulaC9H8O2
InChI KeyILHLUZUMRJQEAH-UHFFFAOYSA-N
SMILESO=C1CC2=CC=CC=C2CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point80°C to 84°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only