p-Toluidine, 99%, crystalline molten mass
p-Toluidine, 99%, crystalline molten mass
p-Toluidine, 99%, crystalline molten mass
Thermo Scientific Chemicals

p-Toluidine, 99%, crystalline molten mass

CAS: 106-49-0 | C7H9N | 107.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
View all
HDPE Bottle
Catalog number 376350050
Price (EUR)
234,00
Each
Quantity:
5 kg
Packaging:
HDPE Bottle
Request bulk or custom format
Price (EUR)
234,00
Each
Chemical Identifiers
CAS106-49-0
IUPAC Name4-methylaniline
Molecular FormulaC7H9N
InChI KeyRZXMPPFPUUCRFN-UHFFFAOYSA-N
SMILESCC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to red
Melting point41°C to 46°C
AppearanceMay discolor to brown on storage
Appearance (Form)Liquid after melting or solid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only