2-Bromobiphenyl, 98%
2-Bromobiphenyl, 98%
2-Bromobiphenyl, 98%
Thermo Scientific Chemicals

2-Bromobiphenyl, 98%

CAS: 2052-07-5 | C12H9Br | 233.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 383450050
Price (EUR)
89,60
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
89,60
Each
Chemical Identifiers
CAS2052-07-5
IUPAC Name2-bromo-1,1'-biphenyl
Molecular FormulaC12H9Br
InChI KeyKTADSLDAUJLZGL-UHFFFAOYSA-N
SMILESBrC1=CC=CC=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.6270 to 1.6310 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only