1-Bromo-5-phenylpentane, 98%
1-Bromo-5-phenylpentane, 98%
1-Bromo-5-phenylpentane, 98%
1-Bromo-5-phenylpentane, 98%
Thermo Scientific Chemicals

1-Bromo-5-phenylpentane, 98%

CAS: 14469-83-1 | C11H15Br | 227.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 384740050
Price (EUR)
256,00
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
256,00
Each
Add to cart
Chemical Identifiers
CAS14469-83-1
IUPAC Name(5-bromopentyl)benzene
Molecular FormulaC11H15Br
InChI KeyQICUPOFVENZWSC-UHFFFAOYSA-N
SMILESBrCCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow to light orange
Infrared spectrumConforms
Refractive index1.5320 to 1.5340 (20°C, 589 nm)
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only