Thermo Scientific Chemicals

4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 397040010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Trifluoromethoxy)phenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 397040010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 397040010
Price (EUR)
Price: 43,10
Online price: 38,36
Your price:
Quantity
-

Chemical Identifiers

CAS
133115-72-7
IUPAC Name
[4-(trifluoromethoxy)phenyl]hydrazine hydrochloride
Molecular Formula
C7H8ClF3N2O
InChI Key
KQXZVSQCMVKMBK-UHFFFAOYSA-N
SMILES
Cl.NNC1=CC=C(OC(F)(F)F)C=C1
Appearance (Color)
White to light brown
Infrared spectrum
Conforms
Titration Argentometric
>=96.0 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water