Albumin, for biochemistry, from chicken egg, powder
Albumin, for biochemistry, from chicken egg, powder
Thermo Scientific Chemicals

Albumin, for biochemistry, from chicken egg, powder

Albumin, bovine serum (BSA), CAS # 9048-46-8, is a small, stable biomolecule derived from bovine blood with wide applications in biochemistry, immunochemistry, pharmacokinetics, and cell culture studies.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 400450025
Price (EUR)
574,00
Each
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (EUR)
574,00
Each
Add to cart
Chemical Identifiers
CAS9006-59-1
MDL NumberMFCD00130424
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Powder
Protein>=80 %
Water=<8.0 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Albumin, bovine serum (BSA) is available as a protease-free, lyophilized powder that may be stably stored at 2-8°c
• It is comprised of 583 amino acid residues, and has a molecular weight of 66.5 kDa

Applications
• BSA is used as a protein standard in quantification assays, such as the Bradford protein assay
• It is used to stabilize enzymes in restriction digestions in molecular biology experiments
• It is a blocking agent in immunoblots thus increasing signal to noise ratios by quenching non-specific immunochemical reactions
• BSA is a nutritive agent and used as a supplement in cell and microbial culture media

RUO – Research Use Only

General References:

  1. Farwell, A. P.; Dubord-Tomasetti, S. A. Thyroid hormone regulates the expression of laminin in the developing rat cerebellum. Endocrinology. 1999,140 (9), 4221–7.
  2. Kreader, C. A. Relief of amplification inhibition in PCR with bovine serum albumin or T4 gene 32 protein. Applied and Environmental Microbiology. 1996, 62 (3), 1102–6.
  3. Axelsson, I. Characterization of proteins and other macromolecules by agarose gel chromatography. Journal of Chromatography A. 1978, 152 (1), 21–32.