3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%
Thermo Scientific Chemicals

3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%

CAS: 2421-28-5 | C17H6O7 | 322.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 401930025
Price (EUR)
388,00
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
388,00
Each
Add to cart
Chemical Identifiers
CAS2421-28-5
IUPAC Name5-(1,3-dioxo-1,3-dihydro-2-benzofuran-5-carbonyl)-1,3-dihydro-2-benzofuran-1,3-dione
Molecular FormulaC17H6O7
InChI KeyVQVIHDPBMFABCQ-UHFFFAOYSA-N
SMILESO=C(C1=CC=C2C(=O)OC(=O)C2=C1)C1=CC=C2C(=O)OC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to tan
Appearance (Form)Powder
Infrared spectrumConforms
Melting point220°C to 229°C
Titration by coupling>=97.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only