4-Nonylphenol, 99%, mixture of isomers
4-Nonylphenol, 99%, mixture of isomers
4-Nonylphenol, 99%, mixture of isomers
Thermo Scientific Chemicals

4-Nonylphenol, 99%, mixture of isomers

CAS: 84852-15-3 | C15H24O | 220.356 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 416240050
Price (EUR)
581,00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
581,00
Each
Chemical Identifiers
CAS84852-15-3
IUPAC Name4-(7-methyloctyl)phenol
Molecular FormulaC15H24O
InChI KeyJSFITYFUKSFPBZ-UHFFFAOYSA-N
SMILESCC(C)CCCCCCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.05 %
Refractive index1.5105 to 1.5165 (20°C, 589 nm)
Appearance (Form)Clear viscous liquid
Color scale=<50 APHA
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only