Pivaloylacetonitrile, 97%
Pivaloylacetonitrile, 97%
Pivaloylacetonitrile, 97%
Thermo Scientific Chemicals

Pivaloylacetonitrile, 97%

CAS: 59997-51-2 | C7H11NO | 125.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 418021000
Price (EUR)
98,30
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
98,30
Each
Chemical Identifiers
CAS59997-51-2
IUPAC Name4,4-dimethyl-3-oxopentanenitrile
Molecular FormulaC7H11NO
InChI KeyMXZMACXOMZKYHJ-UHFFFAOYSA-N
SMILESCC(C)(C)C(=O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)White to cream
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Melting point66°C to 72°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only