Tetrahydrofurfuryl alcohol, 99+%
Tetrahydrofurfuryl alcohol, 99+%
Tetrahydrofurfuryl alcohol, 99+%
Thermo Scientific Chemicals

Tetrahydrofurfuryl alcohol, 99+%

CAS: 97-99-4 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Catalog number 420442500
Price (EUR)
20,60
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
20,60
Each
Chemical Identifiers
CAS97-99-4
IUPAC Name(oxolan-2-yl)methanol
Molecular FormulaC5H10O2
InChI KeyBSYVTEYKTMYBMK-UHFFFAOYNA-N
SMILESOCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC=<0.1 % Furfuryl alcohol
Infrared spectrumConforms
GC=<1.6 % 1,2-Pentanediol
Refractive index1.4510 to 1.4530 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only