1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide, AcroSeal™, Thermo Scientific Chemicals
1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide, AcroSeal™, Thermo Scientific Chemicals
1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide, AcroSeal™, Thermo Scientific Chemicals
1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Propanephosphonic acid cyclic anhydride, 50 wt.% solution in dimethylformamide, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantityPackaging
429111000100 mLAcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 429111000
Price (EUR)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS68957-94-8
IUPAC Nametripropyl-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-trione
Molecular FormulaC9H21O6P3
InChI KeyPAQZWJGSJMLPMG-UHFFFAOYSA-N
SMILESCCCP1(=O)OP(=O)(CCC)OP(=O)(CCC)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Refractive index1.4500 to 1.4540 (20°C, 589 nm)
Appearance (Form)Solution
Assay>=50 % Solids
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only