3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%
Thermo Scientific Chemicals

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%

CAS: 4568-71-2 | C13H16ClNOS | 269.79 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 440520250
Price (EUR)
75,00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
75,00
Each
Add to cart
Chemical Identifiers
CAS4568-71-2
IUPAC Name3-benzyl-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium chloride
Molecular FormulaC13H16ClNOS
InChI KeyIWSVLBKHBJGMAA-UHFFFAOYSA-M
SMILES[Cl-].CC1=C(CCO)SC=[N+]1CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Elemental Analysis5.06 to 5.32 % N
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point141°C to 147°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only