Sodium fluoride, for biochemistry, Thermo Scientific Chemicals
Sodium fluoride, for biochemistry, Thermo Scientific Chemicals
Sodium fluoride, for biochemistry, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium fluoride, for biochemistry, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
447355000500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 447355000
Price (EUR)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7681-49-4
IUPAC Namesodium fluoride
Molecular FormulaFNa
InChI KeyPUZPDOWCWNUUKD-UHFFFAOYSA-M
SMILES[F-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Heavy metals (as Pb)=<0.003 %
Iron (Fe)=<0.003 %
Chloride (Cl)=<0.005 %
Sulfate (SO4)=<0.03 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only