Sodium bismuthate, ACS reagent
Sodium bismuthate, ACS reagent
Sodium bismuthate, ACS reagent
Thermo Scientific Chemicals

Sodium bismuthate, ACS reagent

CAS: 12232-99-4 | BiNaO3 | 279.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 448131000
Price (EUR)
283,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
283,00
Each
Chemical Identifiers
CAS12232-99-4
IUPAC Namesodium bismuthanoylolate
Molecular FormulaBiNaO3
InChI KeyPNYYBUOBTVHFDN-UHFFFAOYSA-N
SMILES[Na+].[O-][Bi](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Oxidizing efficiency>=99.6 %
Titration with KMnO4>=80.0 %
Chloride (Cl)=<0.002 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only