Methyl hexanoate, 99%
Methyl hexanoate, 99%
Methyl hexanoate, 99%
Thermo Scientific Chemicals

Methyl hexanoate, 99%

CAS: 106-70-7 | C7H14O2 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 449240010
Price (EUR)
117,65
Online exclusive
131,00
Save 13,35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
117,65
Online exclusive
131,00
Save 13,35 (10%)
Each
Chemical Identifiers
CAS106-70-7
IUPAC Namemethyl hexanoate
Molecular FormulaC7H14O2
InChI KeyNUKZAGXMHTUAFE-UHFFFAOYSA-N
SMILESCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Colorless to light yellow
Refractive index1.4040 to 1.4060 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only