3-Nitro-1H-pyrazole, 97%, Thermo Scientific Chemicals
3-Nitro-1H-pyrazole, 97%, Thermo Scientific Chemicals
3-Nitro-1H-pyrazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Nitro-1H-pyrazole, 97%, Thermo Scientific Chemicals

Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 451680050
Price (EUR)
102,00
Each
Request bulk or custom format
Price (EUR)
102,00
Each
Chemical Identifiers
CAS26621-44-3
IUPAC Name5-nitro-1H-pyrazole
Molecular FormulaC3H3N3O2
InChI KeyMZRUFMBFIKGOAL-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White to light brown
Melting point175°C to 178°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only