Product Image
Thermo Scientific Chemicals

Chondroitin sulfate sodium salt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 456280010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS9082-07-9
IUPAC Name(2S,3S,4S,5R,6R)-6-{[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-(sulfooxy)oxan-4-yl]oxy}-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular FormulaC13H21NO15S
InChI KeyKXKPYJOVDUMHGS-OSRGNVMNSA-N
SMILESCC(=O)N[C@H]1[C@H](O)O[C@H](OS(O)(=O)=O)[C@H](O)[C@@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Appearance (Form)Powder
Residue after ign. (sulfated)15 % to 30 %
Loss on drying=<12 % (105°C, 4 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only