Chemical Identifiers
CAS169590-42-5
IUPAC Name4-[5-(4-methylphenyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl]benzene-1-sulfonamide
Molecular FormulaC17H14F3N3O2S
InChI KeyRZEKVGVHFLEQIL-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C1=CC(=NN1C1=CC=C(C=C1)S(N)(=O)=O)C(F)(F)F
View more