Riluzole, 98%
Riluzole, 98%
Riluzole, 98%
Thermo Scientific Chemicals

Riluzole, 98%

CAS: 1744-22-5 | C8H5F3N2OS | 234.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
Catalog number 459360010
Price (EUR)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1744-22-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point113°C to 120°C
Water=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only