Product Image
Thermo Scientific Chemicals

L-Biopterin, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 461660010
Price (EUR)
789,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
789,00
Each
Chemical Identifiers
CAS22150-76-1
IUPAC Name2-amino-6-(1,2-dihydroxypropyl)-1,4-dihydropteridin-4-one
Molecular FormulaC9H11N5O3
InChI KeyLHQIJBMDNUYRAM-UHFFFAOYNA-N
SMILESCC(O)C(O)C1=CN=C2NC(N)=NC(=O)C2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<5 %
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only