Sodium sulfobutyl ether beta-cyclodextrin
Sodium sulfobutyl ether beta-cyclodextrin
Thermo Scientific Chemicals

Sodium sulfobutyl ether beta-cyclodextrin

CAS: 182410-00-0 | xNa·x(C4H9O3S) | 1277.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 463360250
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS182410-00-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Water=<10 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only