Product Image
Thermo Scientific Chemicals

Di-n-butyl carbonate

CAS: 542-52-9 | C9H18O3 | 174.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 463880250
Price (EUR)
199,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
199,00
Each
Chemical Identifiers
CAS542-52-9
IUPAC Namedibutyl carbonate
Molecular FormulaC9H18O3
InChI KeyQLVWOKQMDLQXNN-UHFFFAOYSA-N
SMILESCCCCOC(=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Refractive index1.4100 to 1.4140 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only