Product Image
Thermo Scientific Chemicals

N-Oxalylglycine

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 464712500
Price (EUR)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS5262-39-5
IUPAC Name2-(carboxyformamido)acetic acid
Molecular FormulaC4H5NO5
InChI KeyBIMZLRFONYSTPT-UHFFFAOYSA-N
SMILESOC(=O)CNC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
HPLC>=94 %
Appearance (Form)Solid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only