Product Image
Thermo Scientific Chemicals

Flupentixol dihydrochloride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 464820050
Price (EUR)
920,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
920,00
Each
Chemical Identifiers
CAS2413-38-9
IUPAC Namedihydrogen 2-(4-{3-[(9Z)-2-(trifluoromethyl)-9H-thioxanthen-9-ylidene]propyl}piperazin-1-yl)ethan-1-ol dichloride
Molecular FormulaC23H27Cl2F3N2OS
InChI KeyIOVDQEIIMOZNNA-MHKBYHAFSA-N
SMILES[H+].[H+].[Cl-].[Cl-].OCCN1CCN(CC\C=C2\C3=CC=CC=C3SC3=CC=C(C=C23)C(F)(F)F)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(50 mg/ml water) Clear solution
Appearance (Color)White to yellow
Appearance (Color)Powder
Infrared spectrumConforms
Loss on drying=<2 % (105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only