Product Image
Thermo Scientific Chemicals

Bosutinib

CAS: 380843-75-4 | C26H29Cl2N5O3 | 530.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 465652500
Price (EUR)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS380843-75-4
IUPAC Name4-[(2,4-dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methylpiperazin-1-yl)propoxy]quinoline-3-carbonitrile
Molecular FormulaC26H29Cl2N5O3
InChI KeyUBPYILGKFZZVDX-UHFFFAOYSA-N
SMILESCOC1=CC(NC2=C3C=C(OC)C(OCCCN4CCN(C)CC4)=CC3=NC=C2C#N)=C(Cl)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<0.5 %
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only