Proflavine hydrochloride
Proflavine hydrochloride
Proflavine hydrochloride
Thermo Scientific Chemicals

Proflavine hydrochloride

CAS: 952-23-8 | C26H26Cl2N6O | 509.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465860050
Price (EUR)
37,65
線上優惠
41,30
Save 3,65 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
37,65
線上優惠
41,30
Save 3,65 (9%)
Each
Chemical Identifiers
CAS952-23-8
IUPAC Namedihydrogen bis(acridine-3,6-diamine) hydrate dichloride
Molecular FormulaC26H26Cl2N6O
InChI KeyAPOVFGFHTIKDOX-UHFFFAOYSA-N
SMILES[H+].[H+].O.[Cl-].[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark orange to red-brown
Appearance (Form)Powder
Loss on drying=<4 % (105°C, 1 hr)
Solubility(1 mg/ml in water) Clear solution
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only