Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
Thermo Scientific™
Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityFilteredFit for PurposeUnit Size
908-021PK
also known as 908-021
150 μLSterileFilteredSterile and PCR ReadyPack of 960
906-02150 μLSterileFilteredSterile and PCR ReadyCase of 4800
906-021PK
also known as 906-021
50 μLSterileFilteredSterile and PCR ReadyPack of 960
906-26150 μLSterileNon-filteredSterileCase of 4800
906-261PK
also known as 906-261
50 μLSterileNon-filteredSterilePack of 960
906-26250 μLNon-sterileNon-filteredCertified QualityCase of 4800
907-02180 μLSterileFilteredSterile and PCR ReadyCase of 4800
907-021PK
also known as 907-021
80 μLSterileFilteredSterile and PCR ReadyPack of 960
907-261100 μLSterileNon-filteredSterileCase of 4800
907-261PK
also known as 907-261
100 μLSterileNon-filteredSterilePack of 960
907-262100 μLNon-sterileNon-filteredCertified QualityCase of 4800
907-262PK
also known as 907-262
100 μLNon-sterileNon-filteredCertified QualityPack of 960
908-021150 μLSterileFilteredSterile and PCR ReadyCase of 4800
908-261200 μLSterileNon-filteredSterileCase of 4800
908-261PK
also known as 908-261
200 μLSterileNon-filteredSterilePack of 960
908-262200 μLNon-sterileNon-filteredCertified QualityCase of 4800
908-262PK
also known as 908-262
200 μLNon-sterileNon-filteredCertified QualityPack of 960
Catalog number 908-021PK
also known as 908-021
Price (EUR)
160,00
Pack of 960
Volume (Metric):
150 μL
Sterility:
Sterile
Filtered:
Filtered
Fit for Purpose:
Sterile and PCR Ready
Unit Size:
Pack of 960
Price (EUR)
160,00
Pack of 960
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications
ColorClear
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius
FormatRack
Product LineTecan
SterilitySterile
Tip StyleAutomation
Volume (Metric)150 μL
Unit SizePack of 960