alpha-D-Glucose-1-phosphate disodium salt tetrahydrate, 98%
alpha-D-Glucose-1-phosphate disodium salt tetrahydrate, 98%
alpha-D-Glucose-1-phosphate disodium salt tetrahydrate, 98%
Thermo Scientific Chemicals

alpha-D-Glucose-1-phosphate disodium salt tetrahydrate, 98%

CAS: 56401-20-8 | C6H13Na2O9P | 306.114 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12064.14
also known as A12064-14
Price (EUR)
1 043,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
1 043,00
Each
Chemical Identifiers
CAS56401-20-8
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥ 97.5%
Appearance (Color)White
Water Content (Karl Fischer Titration)≤18.0%
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
alpha-D-Glucose-1-phosphate disodium salt tetra hydrate is a glucose molecule that can be used in cellular catabolism. It is used as an intermediate in chemical and pharmaceutical research.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
RUO – Research Use Only

General References:

  1. H Mulhausen.; J Mendicino. A kinetic study of the effect of alpha-D-galactose, alpha-D-mannose, alpha-D-glucosamine, N-acetyl-alpha-D-glucosamine, and alpha-D-ribose diphosphate on the activity of phosphoglucomutase.J Biol Chem.1970,245(16), 4038-4046.
  2. R.M., et al. Prepared by a modification of the procedure of McCready.J. Am. Chem. Soc., 1944,66560.