2-Chloro-4-(trifluoromethyl)aniline, 98%
2-Chloro-4-(trifluoromethyl)aniline, 98%
2-Chloro-4-(trifluoromethyl)aniline, 98%
Thermo Scientific Chemicals

2-Chloro-4-(trifluoromethyl)aniline, 98%

CAS: 39885-50-2 | C7H5ClF3N | 195.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12242.14
also known as A12242-14
Price (EUR)
135,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
135,00
Each
Chemical Identifiers
CAS39885-50-2
IUPAC Name2-chloro-4-(trifluoromethyl)aniline
Molecular FormulaC7H5ClF3N
InChI KeyMBBUTABXEITVNY-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1Cl)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
Refractive Index1.5000-1.5040 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only