3-Cyano-2-hydroxy-4,6-dimethylpyridine, 98%
3-Cyano-2-hydroxy-4,6-dimethylpyridine, 98%
3-Cyano-2-hydroxy-4,6-dimethylpyridine, 98%
Thermo Scientific Chemicals

3-Cyano-2-hydroxy-4,6-dimethylpyridine, 98%

CAS: 769-28-8 | C8H8N2O | 148.165 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12466.22
also known as A12466-22
Price (EUR)
113,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
113,00
Each
Chemical Identifiers
CAS769-28-8
IUPAC Name4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Molecular FormulaC8H8N2O
InChI KeyOCYMJCILWYHKAU-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(C#N)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only