4,4'-Dichlorobenzophenone, 99%
4,4'-Dichlorobenzophenone, 99%
4,4'-Dichlorobenzophenone, 99%
Thermo Scientific Chemicals

4,4'-Dichlorobenzophenone, 99%

CAS: 90-98-2 | C13H8Cl2O | 251.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12608.36
also known as A12608-36
Price (EUR)
440,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
440,00
Each
Chemical Identifiers
CAS90-98-2
IUPAC Namebis(4-chlorophenyl)methanone
Molecular FormulaC13H8Cl2O
InChI KeyOKISUZLXOYGIFP-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Melting Point (clear melt)142.0-149.0?C
Assay (GC)≥98.5%
It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Soluble in hot acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Avoid strong oxidizing agents.
RUO – Research Use Only

General References:

  1. C. Venkatesan.; T. Jaimol.; P. Moreau.; A. Finiels.; A.V. Ramaswamy.; A.P. Singh. Liquid Phase Selective Benzoylation of Chlorobenzene to 4,4'-Dichlorobenzophenone over Zeolite H-Beta. Catal. Lett.. 2001, 75 (1),119-123.
  2. K. Yoneyama.; F. Matsumura. Reductive metabolism of heptachlor, parathion, 4,4'-dichlorobenzophenone, and carbophenothion by rat liver systems. Pestic. Biochem. Physiol. 1981, 15 (3), 213-221.