Calcium hydroxide, 95%
Calcium hydroxide, 95%
Calcium hydroxide, 95%
Calcium hydroxide, 95%
Thermo Scientific Chemicals

Calcium hydroxide, 95%

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A12650.0I
also known as A12650-0I
Price (EUR)
134,00
Each
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (EUR)
134,00
Each
Add to cart
Chemical Identifiers
CAS1305-62-0
IUPAC Namecalcium dihydroxide
Molecular FormulaCaH2O2
InChI KeyAXCZMVOFGPJBDE-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Acid-base back titration≥94.0 to ≤106.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Calcium hydroxide is used as a flocculant in water and sewage treatment. It is also used in fresh water treatment to raise the pH of the water thereby controlling the corrosion of pipes. It is widely used in the production of calcium stearate. It is an active ingredient in whitewash, mortar and plaster. This compound is also used in the petroleum refining industry for the preparation of additives to oils and in the preparation of dry mixes for painting and decorating. It finds application as an antifungal and antimicrobial preservative for vegetables in storage.

Solubility
Soluble in glycerol and acids. Slightly soluble in water.Insoluble in alcohol.

Notes
Air sensitive. Incompatible with strong acids.Incompatible with maleic anhydride, phosphorous, nitroethane, nitromethane, nitorparaffins, nitropropane and strong acids.
RUO – Research Use Only

General References:

  1. Shokaraneh, A.; Farhad, A. R.; Farhad, N.; Saatchi, M.; Hasheminia, S. M. Antibacterial effect of triantibiotic mixture versus calcium hydroxide in combination with active agents against Enterococcus faecalis biofilm. Dent Mater J 2014, 33 (6), 733-738.
  2. Khor, W. C.; Rabaey, K.; Vervaeren, H. Low temperature calcium hydroxide treatment enhances anaerobic methane production from (extruded) biomass. Bioresour. Technol. 2015, 176, 181-188.