Ethyl indole-3-acetate, 98+%
Ethyl indole-3-acetate, 98+%
Ethyl indole-3-acetate, 98+%
Ethyl indole-3-acetate, 98+%
Thermo Scientific Chemicals

Ethyl indole-3-acetate, 98+%

CAS: 778-82-5 | C12H13NO2 | 203.241 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A13474.06
also known as A13474-06
Price (EUR)
63,70
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
63,70
Each
Add to cart
Chemical Identifiers
CAS778-82-5
IUPAC Nameethyl 2-(1H-indol-3-yl)acetate
Molecular FormulaC12H13NO2
InChI KeyHUDBDWIQSIGUDI-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or pale yellow
FormPowder
Assay (GC)≥98.0%
Melting Point (clear melt)39.0-45.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only