p-Toluidine, 99+%
p-Toluidine, 99+%
p-Toluidine, 99+%
Thermo Scientific Chemicals

p-Toluidine, 99+%

CAS: 106-49-0 | C7H9N | 107.156 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A13698.0I
also known as A13698-0I
Price (EUR)
328,00
Each
Quantity:
5000 g
Request bulk or custom format
Price (EUR)
328,00
Each
Chemical Identifiers
CAS106-49-0
IUPAC Name4-methylaniline
Molecular FormulaC7H9N
InChI KeyRZXMPPFPUUCRFN-UHFFFAOYSA-N
SMILESCC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to cream to yellow to orange to red to brown
FormFused solid or liquid after melting
Assay (GC)≥99.0%
CommentMay discolor to brown on storage
Identification (FTIR)Conforms
View more
p-Toluidine is an intermediate in the production of dyes, organic chemicals and aromatic azo compounds. It serves as a component of accelerators for cyanoacrylate glues. Furthermore, it acts as a bidentate Schiff base ligand through condensation with salicylaldehyde. It reacts with catecholamine to form a dye which is useful for spectrophotometric determination of catecholamine drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluidine is an intermediate in the production of dyes, organic chemicals and aromatic azo compounds. It serves as a component of accelerators for cyanoacrylate glues. Furthermore, it acts as a bidentate Schiff base ligand through condensation with salicylaldehyde. It reacts with catecholamine to form a dye which is useful for spectrophotometric determination of catecholamine drugs.

Solubility
Soluble in ethanol, pyridine, diethyl ether, acetone, carbon tetrachloride, methanol, carbon disulfide, oils and dilute acids.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Singh, K.; Dharaiya, N.; Marangoni, D. G.; Bahadur, P. Dissimilar effects of solubilized p-toluidine on the shape of micelles of differently charged surfactants. Colloids Surf., A 2015, 436, 521-529.
  2. Jiang, G.; Tang, B.; Chen, H.; Liu, Y.; Li, L.; Huang, Q.; Chen, W. Controlled growth of hexagonal Zn2GeO4 nanorods on carbon fibers for photocatalytic oxidation of p-toluidine. RSC Adv. 2015, 5 (33), 25801-25805.