n-Butyl formate, 97%
n-Butyl formate, 97%
n-Butyl formate, 97%
Thermo Scientific Chemicals

n-Butyl formate, 97%

CAS: 592-84-7 | C5H10O2 | 102.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1000 mL
Catalog number A13882.AD
also known as A13882-AD
Price (EUR)
25,10
Each
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
25,10
Each
Chemical Identifiers
CAS592-84-7
IUPAC Namebutyl formate
Molecular FormulaC5H10O2
InChI KeyNMJJFJNHVMGPGM-UHFFFAOYSA-N
SMILESCCCCOC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3870-1.3920 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
FormLiquid
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only