Benzyloxyacetaldehyde diethyl acetal, 98%
Benzyloxyacetaldehyde diethyl acetal, 98%
Benzyloxyacetaldehyde diethyl acetal, 98%
Thermo Scientific Chemicals

Benzyloxyacetaldehyde diethyl acetal, 98%

CAS: 42783-78-8 | C13H20O3 | 224.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A14474.09
also known as A14474-09
Price (EUR)
75,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
75,00
Each
Chemical Identifiers
CAS42783-78-8
IUPAC Name[(2,2-diethoxyethoxy)methyl]benzene
Molecular FormulaC13H20O3
InChI KeyVTYTZCJKJNWMGA-UHFFFAOYSA-N
SMILESCCOC(COCC1=CC=CC=C1)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4770-1.4810 @ 20?C
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only