2-Amino-4-methylthiazole, 98+%
2-Amino-4-methylthiazole, 98+%
2-Amino-4-methylthiazole, 98+%
Thermo Scientific Chemicals

2-Amino-4-methylthiazole, 98+%

CAS: 1603-91-4 | C4H6N2S | 114.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A14847.06
also known as A14847-06
Price (EUR)
21,30
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
21,30
Each
Chemical Identifiers
CAS1603-91-4
IUPAC Name4-methyl-1,3-thiazol-2-amine
Molecular FormulaC4H6N2S
InChI KeyOUQMXTJYCAJLGO-UHFFFAOYSA-N
SMILESCC1=CSC(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow to orange
Assay (GC)≥98.0%
Melting Point (clear melt)40-48°C
FormCrystals or powder or crystalline powder or fused/lumpy solid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only